Learn More
NKG2D/CD314 Antibody (1D11) - Low Endotoxin, Azide and BSA Free, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | NKG2D/CD314 |
| Applications | Immunohistochemistry (Frozen), Functional Assay, CyTOF, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry, Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence |
| Classification | Monoclonal |
| Clone | 1D11 |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Flow Cytometry 1:50-1:200, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, CyTOF-ready, Functional Assay |
| Formulation | Phosphate buffered saline with No Preservative |
| Gene Accession No. | P26718 |
| Gene Alias | CD314, CD314 antigen, D12S2489E, FLJ17759, FLJ75772, Killer cell lectin-like receptor subfamily K member 1, killer cell lectin-like receptor subfamily K, member 1, KLR, NK cell receptor D, NKG2-D, NKG2-D type II integral membrane protein, NKG2-D-activating NK receptor, NKG2DDNA segment on chromosome 12 (unique) 2489 expressed sequence |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.