Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NKG2D/CD314 Antibody (2855A), PE/Atto594, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB1391PEATT594
Description
NKG2D/CD314 Monoclonal antibody specifically detects NKG2D/CD314 in Human samples. It is validated for Flow CytometrySpecifications
NKG2D/CD314 | |
Monoclonal | |
PE/ATTO 594 | |
PBS | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
2855A | |
Flow Cytometry | |
CD314, CD314 antigen, D12S2489E, FLJ17759, FLJ75772, Killer cell lectin-like receptor subfamily K member 1, killer cell lectin-like receptor subfamily K, member 1, KLR, NK cell receptor D, NKG2-D, NKG2-D type II integral membrane protein, NKG2-D-activating NK receptor, NKG2DDNA segment on chromosome 12 (unique) 2489 expressed sequence | |
Mouse myeloma cell line, NS0-derived human NKG2D/CD314, Phe78-Val216, Accession # P26718 | |
0.1 mL | |
Adaptive Immunity, Immunology, Neuroscience, Signal Transduction, Stem Cells | |
22914 | |
Store at 4C in the dark. Do not freeze. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction