Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NKG2D (CD314) Mouse anti-Human, Unconjugated, Clone: 1D11, MilliporeSigma™

Catalog No. MABF103225 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF1032-25 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF1032-25 Supplier MilliporeSigma Supplier No. MABF103225UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-NKG2D (CD314), clone 1D11, Cat. No. MABF1032, is a mouse monoclonal antibody that detects human NKG2-D type II integral membrane protein and has been tested for use in Flow Cytometry, Fluorescence Activated Cell Sorting (FACS), Immunohistochemistry, Immunoprecipitation, ECM Adhesion and Inhibit
TRUSTED_SUSTAINABILITY

Specifications

Antigen NKG2D (CD314)
Applications Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Inhibition Assays
Classification Monoclonal
Clone 1D11
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias NKG2-D type II integral membrane protein;Killer cell lectin-like receptor subfamily K member 1;NK cell receptor D;NKG2-D-activating NK receptor;CD314
Gene Symbols KLRK1;D12S2489E;NKG2D
Host Species Mouse
Immunogen Whole NKL cells (aggressive cell line derived from human natural killer cell leukemia).
Molecular Weight of Antigen 25.27 kDa calculated.
Purification Method Protein G purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Inflammation & Immunology
Primary or Secondary Primary
Test Specificity Clone 1D11 detects NKG2-D type II integral membrane protein in human cells.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Product Type Antibodies
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.