Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ NKG2D/CD314 Recombinant Protein
SDP

Catalog No. NB6021650UG Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
250 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB6021650UG 50 μg
NB60216250U 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB6021650UG Supplier Novus Biologicals™ Supplier No. NBP26021650UG

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A recombinant protein with a C-Terminal hIgG-His-tag and corresponding to the amino acids 73-216 of Human NKG2D/CD314 The Recombinant Human NKG2D/CD314 Protein is derived from Baculovirus. The Recombinant Human NKG2D/CD314 Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration 0.5mg/mL
For Use With (Application) SDS-PAGE
Formulation PBS (pH 7.4) containing 10% glycerol. containingno Preservative
Gene ID (Entrez) 22914
Name Recombinant NKG2D/CD314 Protein
Purification Method >95%, by SDS-PAGE
Quantity 50 μg
Source Baculovirus
Storage Requirements Store at 4°Cshort term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Endotoxin Concentration < 1.0 EU per 1ug of protein (determined by LAL method)
Gene Alias CD314, CD314 antigen, D12S2489E, FLJ17759, FLJ75772, Killer cell lectin-like receptor subfamily K member 1, killer cell lectin-like receptor subfamily K, member 1, KLR, NK cell receptor D, NKG2-D, NKG2-D type II integral membrane protein, NKG2-D-activatin
Gene Symbol KLRK1
Product Type Recombinant Protein
Recombinant Recombinant
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.