Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NKG2E Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA580761
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA580761 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA580761 Supplier Invitrogen™ Supplier No. PA580761
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human KLRC3/NKG2E.

Natural killer (NK) cells are lymphocytes that can mediate lysis of certain tumor cells and virus-infected cells without previous activation. They can also regulate specific humoral and cell-mediated immunity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NKG2E
Applications Western Blot
Classification Polyclonal
Concentration 5 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene KLRC3
Gene Accession No. Q07444
Gene Alias killer cell lectin like receptor C3; killer cell lectin-like receptor subfamily C, member 2; killer cell lectin-like receptor subfamily C, member 3; Klrc2; KLRC3; natural killer cell group 2E cell receptor; NK cell receptor E; NKG2E; NKG2-E; NKG2-E type II integral membrane protein; NKG2-E-activating NK receptor
Gene Symbols KLRC3
Host Species Rabbit
Immunogen A synthetic peptide corresponding to the N-terminus of the Human KLRC3/NKG2E.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3823
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.