Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NKG7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144131
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144131 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144131 Supplier Invitrogen™ Supplier No. PA5144131
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NKG7 Polyclonal Antibody for ICC/IF, ELISA

NKG7 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NKG7
Applications ELISA, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene NKG7
Gene Accession No. Q16617
Gene Alias G-CSF-induced gene 1 protein; GIG1; GIG-1 protein; GMP-17; granule membrane protein 17; granule membrane protein of 17 kDa; natural killer cell granule protein 7; natural killer cell group 7 sequence; natural killer cell protein 7; NKG7; p15-TIA-1; Protein NKG7
Gene Symbols NKG7
Host Species Rabbit
Immunogen Recombinant Human Protein NKG7 protein (30-60AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4818
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.