Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NKIRAS1 Monoclonal Antibody (OTI8B11), TrueMAB™, OriGene
SDP

Catalog No. 502539666
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-253-9666 30 μL
50-167-5963 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-253-9666 Supplier OriGene Supplier No. TA505497S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NKIRAS1 Monoclonal antibody specifically detects NKIRAS1 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen NKIRAS1
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI8B11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene RCAN3
Gene Accession No. Q9UKA8
Gene Alias 2400004O09Rik; C15orf1; I1PP2A; I-kappa-B-interacting Ras-like protein 1; kappa B-ras 1; kappa B-Ras protein 1; kappaB-Ras1; KBRAS1; LANP; MAPM; MGC119787; MGC150373; NF-kappa-B inhibitor-interacting Ras-like protein 1; NFKB inhibitor interacting Ras like 1; NFKB inhibitor interacting Ras-like 1; NFKB inhibitor interacting Ras-like 1 S homeolog; NFKB inhibitor interacting Ras-like protein 1; Nkiras1; nkiras1.S; PHAP1; PHAPI; PP32; XELAEV_18033117mg
Host Species Mouse
Immunogen Full length human recombinant protein of human RCAN3 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11123
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.