Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NKp30/NCR3 Antibody (MM0490-9D31), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP211819UV
Description
NKp30/NCR3 Monoclonal specifically detects NKp30/NCR3 in Human samples. It is validated for Flow Cytometry.Specifications
NKp30/NCR3 | |
Monoclonal | |
DyLight 350 | |
Q05D23 | |
NCR3 | |
Human recombinant NKp30/NCR3 | |
0.1 mL | |
Adaptive Immunity, Checkpoint signaling, Immunology, Stem Cells | |
259197 | |
Store at 4C in the dark. | |
IgG2 |
Flow Cytometry | |
MM0490-9D31 | |
Flow Cytometry | |
Activating natural killer receptor p30, CD337 antigen, CD337activating NK-A1 receptor, LY117,1C7NK-p30, lymphocyte antigen 117, natural cytotoxicity triggering receptor 3, Natural killer cell p30-related protein, NKP30, NKp30MALS | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction