Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NKp30 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596595
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596595 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596595 Supplier Invitrogen™ Supplier No. PA596595
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: Rat liver; Cellular Location: Cell membrane, Single-pass type I membrane protein Immunogen sequence: LWVSQPPEIR TLEGSSAFLP CSFNASQGRL AIGSVTWFRD EVVPGKEVRN GTPEFRGRLA PLASSRFLHD HQAELHIRDV RGHDASIYVC RVEVLGLGVG TGNGTRLVVE KEHPQLG.

The protein encoded by this gene is a natural cytotoxicity receptor (NCR) that may aid NK cells in the lysis of tumor cells. The encoded protein interacts with CD3-zeta (CD247), a T-cell receptor. A single nucleotide polymorphism in the 5' untranslated region of this gene has been associated with mild malaria suceptibility. Three transcript variants encoding different isoforms have been found for this gene.[provided by RefSeq, May 2010].
TRUSTED_SUSTAINABILITY

Specifications

Antigen NKp30
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.59 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Ncr3
Gene Accession No. O14931, Q8CFD9
Gene Alias 1C7; Activating natural killer receptor p30; activating NK-A1 receptor; CD337; LY117; lymphocyte antigen 117; MALS; Natural cytotoxicity triggering receptor 3; natural killer cell p30-related protein; natural killer cell receptor; Ncr3; NK receptor 1c7; Nkp30; NK-p30; NKp30S; NKp30v1; NKp30v1ED
Gene Symbols Ncr3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 19-135 of human NCR3 (NP_6673411).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 259197, 294251
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.