Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NKp44 (CD336) Mouse anti-Human, RB780, Clone: p44-8, BD Horizon™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 569203
Description
NKp44 (CD336) Monoclonal antibody specifically detects NKp44 (CD336) in Human samples. It is validated for Flow cytometry
Specifications
NKp44 (CD336) | |
Monoclonal | |
RB780 | |
CD336; NCR2; NCTR2; NKp44; NK-p44; LY95; dJ149M18.1 | |
Human NKp44 | |
100 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unreacted dye was removed. | |
IgG1 κ |
Flow Cytometry | |
p44-8 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Chromatography | |
RUO | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction