Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NKp44 (CD336) Mouse anti-Human, Unconjugated, Clone: 3.43.13, MilliporeSigma™
Mouse Monoclonal Antibody
$189.29 - $477.80
Specifications
| Antigen | NKp44 (CD336) |
|---|---|
| Clone | 3.43.13 |
| Applications | Flow Cytometry, Immunocytochemistry |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
MABF22502MI
|
MilliporeSigma
MABF225025UL |
25 μL |
Each for $189.29
|
|
|||||
|
MABF22501MI
|
MilliporeSigma
MABF2250100UL |
100 μL |
Each for $477.80
|
|
|||||
Description
NKp44 (CD336) Monoclonal antibody specifically detects NKp44 (CD336) in Human samples. It is validated for Flow Cytometry, Affects Function, Immunocytochemistry.Specifications
| NKp44 (CD336) | |
| Flow Cytometry, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| Natural cytotoxicity triggering receptor 2;Lymphocyte antigen 95 homolog;NK cell-activating receptor;Natural killer cell p44-related protein;NK-p44;NKp44;CD336 | |
| NKp44-human crystallizable fragment (Fc) fusion protein. | |
| Primary | |
| Stable for 1 year at -20°C from date of receipt. | |
| 30.68 kDa calculated. |
| 3.43.13 | |
| Monoclonal | |
| Purified | |
| Inflammation and Immunology | |
| NP_004819 | |
| NCR2, LY95 | |
| IgG1 κ | |
| Protein G Purified | |
| Clone 3.43.13 is a mouse monoclonal antibody that detects human Natural cytotoxicity triggering receptor 2 (NKp44) protein. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title