Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NKp44/NCR2 Antibody (1G6), Alexa Fluor™ 700, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP242683AF700

Catalog No. NBP242683AI


Only null left
Add to Cart

Description

Description

NKp44/NCR2 Monoclonal specifically detects NKp44/NCR2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

NKp44/NCR2
Monoclonal
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
NCR2
Recombinant human NKp44 (19-130aa) purified from E. coli
100 μL
Innate Immunity
9436
Store at 4C in the dark.
IgG2a κ
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
1G6
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
CD336, CD336 antigen, LY95lymphocyte antigen 95 homolog (activating NK-receptor; NK-p44), lymphocyte antigen 95 (activating NK-receptor; NK-p44), Lymphocyte antigen 95 homolog, natural cytotoxicity triggering receptor 2, Natural killer cell p44-related protein, NK cell activating receptor (NKp44), NK cell-activating receptor, NKP44, NK-p44dJ149M18.1
Mouse
Protein A purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.