Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NKp44 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103955
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103955 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103955 Supplier Thermo Scientific Supplier No. PA5103955
Add to Cart
Add to Cart

NKp44 Polyclonal Antibody

NKp44 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen NKp44
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene NCR2
Gene Accession No. O95944
Gene Alias CD336; CD336 antigen; dJ149M18.1; LY95; lymphocyte antigen 95 (activating NK-receptor; NK-p44); Lymphocyte antigen 95 homolog; lymphocyte antigen 95 homolog (activating NK-receptor; NK-p44); natural cytotoxicity triggering receptor 2; Natural killer cell p44-related protein; natural killer cell receptor; NCR2; NK cell activating receptor (NKp44); NK cell-activating receptor; NKP44; NK-p44; NK-p44dJ149M18.1
Gene Symbols NCR2
Host Species Rabbit
Immunogen A synthesized peptide derived from human NCR2(Accession O95944), corresponding to amino acid residues T136-G186.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9436
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.