Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NKp44 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Supplier: Thermo Scientific PA547279
Description
In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human (rh) NKp30, rhNKp46, and rhNKp80 is observed. Reconstitute at 0.2 mg/mL in sterile PBS. Endoxin level is <1.5 EU per 1 μg of the antibody by the LAL method.
NKp44 Polyclonal antibody specifically detects NKp44 in Human samples. It is validated for Functional Assay, Western Blot.
Specifications
NKp44 | |
Polyclonal | |
Unconjugated | |
NCR2 | |
CD336; CD336 antigen; dJ149M18.1; LY95; lymphocyte antigen 95 (activating NK-receptor; NK-p44); Lymphocyte antigen 95 homolog; lymphocyte antigen 95 homolog (activating NK-receptor; NK-p44); natural cytotoxicity triggering receptor 2; Natural killer cell p44-related protein; natural killer cell receptor; NCR2; NK cell activating receptor (NKp44); NK cell-activating receptor; NKP44; NK-p44; NK-p44dJ149M18.1 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
9436 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Functional Assay, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
O95944 | |
NCR2 | |
Mouse myeloma cell line NS0-derived recombinant human NKp44/NCR2 Gln22-Pro190, predicted | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction