Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NKp46 (CD335), Mouse anti-Human, Clone: 461-G1, Millipore Sigma™
Mouse Monoclonal Antibody
$207.00 - $523.50
Specifications
Antigen | NKp46 (CD335) |
---|---|
Clone | 461-G1 |
Applications | ELISA, Flow Cytometry |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABF205925
|
MilliporeSigma
MABF205925UG |
25 μg |
Each for $207.00
|
|
|||||
MABF2059100
|
MilliporeSigma
MABF2059100UG |
100 μg |
Each for $523.50
|
|
|||||
Description
Anti-NKp46 (CD335), clone 461-G1, Cat. No. MABF2059, is a mouse monoclonal antibody that detects Natural cytotoxicity triggering receptor 1 and has been tested for use in ELISA and Flow Cytometry.
Natural cytotoxicity triggering receptor 1 (UniProt: O76036; also known as Lymphocyte antigen 94 homolog, NK cell-activating receptor, Natural killer cell p46-related protein, NK-p46, NKp46, hNKp46, CD335) is encoded by the NCR1 (also known as LY94) gene (Gene ID: 9437) in human. NK-p46 is a single-pass type I membrane protein that belongs to the natural cytotoxicity receptor (NCR) family and serves as a cytotoxicity-activating receptor that may contribute to the increased efficiency of activated natural killer (NK) cells to mediate tumor cell lysis and elimination of virus-infected cells. It is considered as a primary activating receptor mediating NK cell antiviral function. It is involved in the functional recognition of viral HA and hemagglutinin-neuraminidases. NKp46 is shown to be predominantly expressed in the nasal lavage of patients infected with respiratory viruses, but not in normal healthy subjects. Following the initiation of an NK-target cell interaction, NKp46 is reported to cluster at the cell membrane, specifically at the immune synapse where it mediates cytoskeletal rearrangement and cellular polarization. Human NK-p46 is synthesized with a signal peptide of 21 amino acids, which is subsequently cleaved off. The mature form has an extracellular domain (aa 22-258), a helical domain (aa 259-279), and a cytoplasmic domain (aa 280-304). It also has two Ig-like domains (aa 34-118 and 129-211) termed as D1 and D2. Five different isoforms of NKp46 have been described that are produced by alternative splicing. (Ref.: Shemer-Avni, Y., et al., (2017). Front. Immunol. 8 (Article 161); 1-10).Specifications
NKp46 (CD335) | |
ELISA, Flow Cytometry | |
Unconjugated | |
Mouse | |
Inflammation & Immunology | |
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. | |
NP_001138929 | |
Human IgG1 Fc-fused to a recombinant fragment corresponding to the first 100 amino acids from the extracellular domain of human NKp46. | |
Primary | |
Stable for 1 year at 2-8°C from date of receipt. |
461-G1 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Natural cytotoxicity triggering receptor 1;Lymphocyte antigen 94 homolog;NK cell-activating receptor;Natural killer cell p46-related protein;NK-p46;hNKp46;CD335 | |
NCR1;LY94 | |
IgG1 κ | |
Protein G purified | |
Clone 461-G1 specifically recognizes whole human NKp46 and NKp46-D1, but does not recognize NKp46-D2. It targets an epitope with in the first 100 amino acids from the extracellular domain. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title