Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NKp46/NCR1 Mouse anti-Human, Clone: B-L46, Novus Biologicals™

Mouse Monoclonal Antibody
$400.50 - $771.00
Specifications
| Antigen | NKp46/NCR1 |
|---|---|
| Clone | B-L46 |
| Concentration | 1 mg/mL |
| Dilution | Flow Cytometry, Functional |
| Applications | Flow Cytometry, Functional Assay |
Description
NKp46/NCR1 Monoclonal antibody specifically detects NKp46/NCR1 in Human samples. It is validated for Flow Cytometry, Functional AssaySpecifications
| NKp46/NCR1 | |
| 1 mg/mL | |
| Flow Cytometry, Functional Assay | |
| Unconjugated | |
| Mouse | |
| Adaptive Immunity, Immunology, Signal Transduction | |
| PBS. Sterile-filtered through 0.22 um. | |
| 9437 | |
| IgG2a κ | |
| Affinity purified | |
| Antibody |
| B-L46 | |
| Flow Cytometry, Functional | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD335, CD335 antigen, hNKp46, LY94lymphocyte antigen 94 homolog (activating NK-receptor; NK-p46), Lymphocyte antigen 94 homolog, natural cytotoxicity triggering receptor 1, Natural killer cell p46-related protein, NK cell-activating receptor, NKp46, NKP46FLJ99094, NK-p46lymphocyte antigen 94 (mouse) homolog (activating NK-receptor; NK-p46) | |
| Recombinant human NKp46/NCR1 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title