Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NKp80 Mouse anti-Human, Clone: 5D12, MilliporeSigma™

Catalog No. MABF2223100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF2223100 100 μg
MABF222325 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF2223100 Supplier MilliporeSigma Supplier No. MABF2223100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NKp80 Monoclonal antibody specifically detects NKp80 in Human samples. It is validated for Flow Cytometry, Immunoprecipitation, Inhibition.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NKp80
Applications Flow Cytometry, Immunoprecipitation, Inhibition Assays
Classification Monoclonal
Clone 5D12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG1 in buffer containing PBS without preservatives.
Host Species Mouse
Immunogen Recombinant extracellular domain of human NKp80.
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Stable for 1 year at +2°C to +8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.