Learn More
Description
Specifications
Specifications
| Antigen | NKp80 |
| Applications | Flow Cytometry, Immunoprecipitation, Inhibition Assays |
| Classification | Monoclonal |
| Clone | 5D12 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | Purified mouse monoclonal antibody IgG1 in buffer containing PBS without preservatives. |
| Host Species | Mouse |
| Immunogen | Recombinant extracellular domain of human NKp80. |
| Purification Method | Protein G purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.