Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NKR-P1B (CD161b) Mouse anti-Mouse, Unconjugated, Clone: 2D12, MilliporeSigma™

Catalog No. MABF21152MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF21152MI 25 μL
MABF21151MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF21152MI Supplier MilliporeSigma Supplier No. MABF211525UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NKR-P1B (CD161b) Monoclonal antibody specifically detects NKR-P1B (CD161b) in Mouse samples. It is validated for Flow Cytometry, Function Assay, Immunoprecipitation, Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NKR-P1B (CD161b)
Applications Flow Cytometry, Functional Assay, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 2D12
Conjugate Unconjugated
Gene Accession No. NP_085102.4
Gene Alias Killer cell lectin-like receptor subfamily B member 1B allele B;CD161 antigen-like family member B;Inhibitory receptor NKR-P1B;Lymphocyte antigen 55d;Ly-55d;Natural killer cell surface protein NKR-P1B allele B6;CD161b
Gene Symbols Klrb1b, Klrb1d, Ly55d, Nkrp1b, Nkrp1d
Host Species Mouse
Immunogen Mouse BaF/3 cells (murine IL-3-dependent pro-B cell line) expressing mouse NKR-P1B protein.
Molecular Weight of Antigen ∼52 kDa observed; 25.03 kDa calculated. . Uncharacterized bands may be observed in some lysate(s).
Purification Method Protein G Purified
Quantity 25 μL
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Test Specificity Clone 2D12 is a mouse monoclonal antibody that detects Natural killer cell surface protein NKR-P1B allele B6 (NKR-P1B) in murine species. It does not react with Nkrp1a, Nkrp1c, and Nkrp1f.
Target Species Mouse
Content And Storage Stable for 1 year at -20°C from date of receipt.
Form Purified
Isotype IgG2a λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.