Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NKR-P1B (CD161b) Rat anti-Rat, MilliporeSigma™

Catalog No. MABF197125 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF1971-25 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF1971-25 Supplier MilliporeSigma Supplier No. MABF197125UL
Add to Cart
Add to Cart

Rat Monoclonal Antibody

This rat monoclonal Anti-NKR-P1B (CD161b), clone Stok27 Antibody for the detection of NKR-P1B.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NKR-P1B (CD161b)
Applications Flow Cytometry, Immunoprecipitation
Classification Monoclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols Klrb1b,Nkrp1b,Nkrp1c
Host Species Rat
Immunogen IL-2-activated NK cells from PVG.1N (RT1n) rats.
Purification Method Protein G Purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Test Specificity Clone Stok27 is a rat monoclonal antibody that detects Killer cell lectin-like receptor subfamily B member 1B allele B (NKR-P1B) in rat. It targets an epitope within the extracellular domain.
Target Species Rat
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.