Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NKR-P1D Antibody (4E9), Alexa Fluor™ 594, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP250531AF594
Description
NKR-P1D Monoclonal specifically detects NKR-P1D in Mouse samples. It is validated for Flow Cytometry.Specifications
NKR-P1D | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
YB2/0 rat plasmacytoma cells transfected with NKRP1 | |
0.1 mL | |
Mouse | |
IgG2b |
Flow Cytometry | |
4000000000 | |
Flow Cytometry | |
Rat | |
Protein G purified | |
Primary | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction