Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NKR-P1D Antibody (4E9) - BSA Free, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP2505310.025ML
Description
NKR-P1D Monoclonal antibody specifically detects NKR-P1D in Mouse samples. It is validated for Functional AssaySpecifications
NKR-P1D | |
Monoclonal | |
1 mg/mL | |
PBS | |
Rat | |
Protein G purified | |
RUO | |
Mouse | |
Purified |
Functional Assay | |
4E9 | |
Unconjugated | |
AI462337, CD161, Klrb1b, Klrb1c, Klrb1d, Ly55b, Ly55c, ly-55c, Ly55d, Ly59, Ly-59, Nk1, Nk-1, NK1.1, Nk1.2, Nk-1.2, NKRP1, NK-RP1, NKR-P1.9, NKRP140, Nkrp1b, Nkrp1-b, NKR-P1B, Nkrp1c, Nkrp1d, NKR-P1D | |
YB2/0 rat plasmacytoma cells transfected with NKR-P1D | |
0.025 mL | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction