Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NKRF Antibody (OTI4C2), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273001AF532
Description
NKRF Monoclonal specifically detects NKRF in Human, Mouse samples. It is validated for Western Blot.Specifications
NKRF | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
NKRF | |
Recombinant protein fragment corresponding to amino acids 1-296 of human NKRF (NP_060014) produced in E.coli. | |
0.1 mL | |
55922.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI4C2 | |
Western Blot | |
ITBA4NFkB-repressing factor, NF-kappaB repressing factor, NFKB repressing factor, NRFNF-kappa-B-repressing factor, Protein ITBA4, Transcription factor NRF | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction