Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NKRF Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117006
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117006 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117006 Supplier Invitrogen™ Supplier No. PA5117006
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: U-87MG, A-549, Mouse brain, Mouse spleen, Mouse kidney, Rat brain Immunogen sequence: YGTKKTSKHA AADEALKILQ KTQPTYPSVK SSQCHTGSSP RGSGKKKDIK DLVVYENSSN PVCTLNDTAQ FNRMTVEYVY ERMTGLRWKC KVILESEVIA EAVGVKKTVK YEAAGEAVKT LKKTQPTVIN NLKKGAVEDV ISRNEIQGRS AEEAYKQQIK EDNIGNQLLR KMGWTGGGLG KSGEGIREPI SVKEQHKREG LGLDVERVNK IAKRDIEQII RNYARSESHT DLTFSRELTN DERKQIHQIA QKYGLKSKSH GVGHDRYLVV GRKRRKEDLL DQLKQEGQVG HYELVMPQAN.

This gene encodes a transcription factor that interacts with specific negative regulatory elements (NREs) to mediate transcriptional repression of certain NK-kappa-B-responsive genes. The protein localizes predominantly to the nucleolus with a small fraction found in the nucleoplasm and cytoplasm.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NKRF
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.81 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene NKRF
Gene Accession No. O15226, Q8BY02
Gene Alias 9430034D17Rik; AW124770; C80287; ITBA4; ITBA4 protein; LOC100910054; NF-kappaB repressing factor; NF-kappa-B-repressing factor; NF-kappa-B-repressing factor-like; NFKB repressing factor; NFkB-repressing factor; NKRF; NRF; Protein ITBA4; Transcription factor NRF
Gene Symbols NKRF
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 391-690 of human NKRF (NP_0600142).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100910054, 55922, 77286
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.