Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NKX1-2 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299300JF646
Description
NKX1-2 Polyclonal antibody specifically detects NKX1-2 in Mouse samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
NKX1-2 | |
Polyclonal | |
Western Blot, Immunoprecipitation | |
bB238F13.2, C10orf121, NKX-1.1, NKX1-2 NK1 homeobox 2 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Mouse NKX1-2. | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Western Blot, Immunoprecipitation | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
390010 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction