Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Nkx1.2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA581228
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA581228 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA581228 Supplier Invitrogen™ Supplier No. PA581228
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody has specificity for Mouse NKX1-2.

NKX1-2 may function in cell specification, particularly in the CNS.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nkx1.2
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.03% ProClin 300; pH 7
Gene NKX1-2
Gene Accession No. P42580
Gene Alias bB238F13.2; C10orf121; Homeobox protein SAX-1; NK1 homeobox 2; NK1 transcription factor related, locus 2; NK1 transcription factor related, locus 2 (Drosophila); NK1 transcription factor-related protein 2; Nkx1.1; NKX-1.1; Nkx-1.2; NKX11; NKX-11; NKX1-2; Sax1; spinal cord axial homeobox gene 1
Gene Symbols NKX1-2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to the N-terminus of the Mouse NKX1-2.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20231
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.