Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nkx1.2 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA556121
Description
Immunogen sequence: TYEQLVALEN KFRATRYLSV CERLNLALSL SLTETQVK Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
Specifications
| Nkx1.2 | |
| Polyclonal | |
| Unconjugated | |
| NKX1-2 | |
| NKX1-2 | |
| Recombit protein corresponding to Human Nkx1.2 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UD57 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 390010 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Research Use Only
Spot an opportunity for improvement?Share a Content Correction