Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Nkx2.1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102693
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102693 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102693 Supplier Invitrogen™ Supplier No. PA5102693
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total TTF1.

This gene encodes a protein initially identified as a thyroid-specific transcription factor. The encoded protein binds to thyroglobulin promoter and regulates the expression of thyroid-specific genes but has also been shown to regulate the expression of genes involved in morphogenesis. Mutations and deletions in this gene are associated with benign hereditary chorea, choreoathetosis, congenital hypothyroidism, and neonatal respiratory distress, and may be associated with thyroid cancer. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nkx2.1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene NKX2-1
Gene Accession No. P23441, P43699, P50220
Gene Alias AV026640; BCH; BHC; homeobox protein NK-2 homolog A; Homeobox protein Nkx-2.1; NK-2; NK2 homeobox 1; NK-2 homolog A; NKX2.1; Nkx-2.1; NKX2-1; NKX2A; NMTC1; RNA polymerase I transcription termination factor; T/EBP; TEBP; Thyroid nuclear factor 1; thyroid transcription factor 1; thyroid transcription factor 1 TTF-1 NK-2; thyroid transcription factor 1-like; thyroid-specific enhancer-binding protein; tinman; TITF1; TTF1; TTF-1
Gene Symbols NKX2-1
Host Species Rabbit
Immunogen A synthesized peptide derived from human NKX2-1(Accession P43699), corresponding to amino acid residues M1-P49.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21869, 25628, 7080
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.