Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NKX2.2 Antibody (rNX2/1523), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308567AF405
Description
NKX2.2 Monoclonal antibody specifically detects NKX2.2 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
NKX2.2 | |
Monoclonal | |
Alexa Fluor 405 | |
Homeobox protein NK-2 homolog B, NK-2 (Drosophila) homolog B, NK2 homeobox 2, NK2 transcription factor related, locus 2, NK2 transcription factor related, locus 2 (Drosophila), NK2 transcription factor-like protein B, Nkx2.2, NKX2.2homeobox protein Nkx-2.2, NKX2BNK-2 homolog B | |
Human recombinant NKX2.2 protein fragment (around aa1-119) (exact sequence is proprietary) (Uniprot: O95096) | |
100 μg | |
Primary | |
Human | |
Purified |
Immunohistochemistry (Paraffin) | |
rNX2/1523 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
4821 | |
Store at 4°C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction