Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Nkx2.2 Monoclonal Antibody (3E4)
GREENER_CHOICE

Catalog No. PIMA517138
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA517138 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA517138 Supplier Invitrogen™ Supplier No. MA517138
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-17138 targets NKX2.2 in FACS, indirect ELISA, and WB applications and shows reactivity with Human samples. The MA5-17138 immunogen is purified recombinant fragment of human NKX2.2 expressed in E. Coli. MA5-17138 detects NKX2.2 which has a predicted molecular weight of approximately 30.1kDa.

The protein encoded by this gene contains a homeobox domain and may be involved in the morphogenesis of the central nervous system. This gene is found on chromosome 20 near NKX2-4, and these two genes appear to be duplicated on chromosome 14 in the form of TITF1 and NKX2-8. The encoded protein is likely to be a nuclear transcription factor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nkx2.2
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 3E4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene NKX2-2
Gene Accession No. O95096
Gene Alias Drosophila NK2 transcription factor related, locus 2; glycoprotein GP330, renal; homeobox protein NK-2 homolog B; Homeobox protein Nkx-2.2; homeobox protein Nkx2-2; NK2 homeobox 2; NK2 transcription factor related locus 2; NK2 transcription factor related, locus 2; NK2 transcription factor-like protein B; NKX2.2; Nkx-2.2; Nkx2\.2; Nkx2-2; NKX2B; tinman
Gene Symbols NKX2-2
Host Species Mouse
Immunogen Purified recombinant fragment of human NKX2.2 expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4821
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.