Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NKX2.2 Antibody (NX2/1524), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP254323AF532
Description
NKX2.2 Monoclonal specifically detects NKX2.2 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
NKX2.2 | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
NKX2-2 | |
Recombinant fragment (around aa1-119) of human NKX2.2 protein (exact sequence is proprietary) (Uniprot: O95096) | |
0.1 mL | |
4821.0 | |
Store at 4°C in the dark. |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
NX2/1524 | |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
Homeobox protein NK-2 homolog B, NK-2 (Drosophila) homolog B, NK2 homeobox 2, NK2 transcription factor related, locus 2, NK2 transcription factor related, locus 2 (Drosophila), NK2 transcription factor-like protein B, Nkx2.2, NKX2.2homeobox protein Nkx-2.2, NKX2BNK-2 homolog B | |
Mouse | |
Protein A or G purified | |
Primary | |
Human | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction