Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Nkx2.2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572761
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572761 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572761 Supplier Invitrogen™ Supplier No. PA572761
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Nkx2.2 Polyclonal Antibody for Western Blot, IHC (P), ELISA

The protein encoded by this gene contains a homeobox domain and may be involved in the morphogenesis of the central nervous system. This gene is found on chromosome 20 near NKX2-4, and these two genes appear to be duplicated on chromosome 14 in the form of TITF1 and NKX2-8. The encoded protein is likely to be a nuclear transcription factor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nkx2.2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene NKX2-2
Gene Accession No. O95096, P42586
Gene Alias Drosophila NK2 transcription factor related, locus 2; glycoprotein GP330, renal; homeobox protein NK-2 homolog B; Homeobox protein Nkx-2.2; homeobox protein Nkx2-2; NK2 homeobox 2; NK2 transcription factor related locus 2; NK2 transcription factor related, locus 2; NK2 transcription factor-like protein B; NKX2.2; Nkx-2.2; Nkx2\.2; Nkx2-2; NKX2B; tinman
Gene Symbols NKX2-2
Host Species Rabbit
Immunogen NKX2-2 antibody for a 19 amino acid synthetic peptide near the center of human NKX2-2. The immunogen is within amino acids 90 - 140 of NKX2-2.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18088, 366214, 4821
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.