Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Nkx2.2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578079
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578079 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578079 Supplier Invitrogen™ Supplier No. PA578079
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: Nkx2.2-transfected 293T Predicted Reactivity: Mouse (100%), Chicken (89%), Bovine (94%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The protein encoded by this gene contains a homeobox domain and may be involved in the morphogenesis of the central nervous system. This gene is found on chromosome 20 near NKX2-4, and these two genes appear to be duplicated on chromosome 14 in the form of TITF1 and NKX2-8. The encoded protein is likely to be a nuclear transcription factor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nkx2.2
Applications Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.13 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene NKX2-2
Gene Accession No. P42586
Gene Alias Drosophila NK2 transcription factor related, locus 2; glycoprotein GP330, renal; homeobox protein NK-2 homolog B; Homeobox protein Nkx-2.2; homeobox protein Nkx2-2; NK2 homeobox 2; NK2 transcription factor related locus 2; NK2 transcription factor related, locus 2; NK2 transcription factor-like protein B; NKX2.2; Nkx-2.2; Nkx2\.2; Nkx2-2; NKX2B; tinman
Gene Symbols NKX2-2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of mouse Nkx2.2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18088, 366214
Target Species Mouse, Rat, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.