Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Nkx2.2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5115656
Description
Antibody detects endogenous levels of total NKX22.
The protein encoded by this gene contains a homeobox domain and may be involved in the morphogenesis of the central nervous system. This gene is found on chromosome 20 near NKX2-4, and these two genes appear to be duplicated on chromosome 14 in the form of TITF1 and NKX2-8. The encoded protein is likely to be a nuclear transcription factor.
Specifications
Nkx2.2 | |
Polyclonal | |
Unconjugated | |
NKX2-2 | |
Drosophila NK2 transcription factor related, locus 2; glycoprotein GP330, renal; homeobox protein NK-2 homolog B; Homeobox protein Nkx-2.2; homeobox protein Nkx2-2; NK2 homeobox 2; NK2 transcription factor related locus 2; NK2 transcription factor related, locus 2; NK2 transcription factor-like protein B; NKX2.2; Nkx-2.2; Nkx2\.2; Nkx2-2; NKX2B; tinman | |
Rabbit | |
Affinity chromatography | |
RUO | |
18088, 366214, 4821 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide | |
O95096, P42586 | |
NKX2-2 | |
A synthesized peptide derived from human NKX2-2(Accession O95096), corresponding to amino acid residues H88-A138. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction