Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NKX2-4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP25785425UL
Description
NKX2-4 Polyclonal specifically detects NKX2-4 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
NKX2-4 | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence 1-4 μg/mL | |
homeobox protein Nkx-2.4, NK2 homeobox 4, NKX2.4, NKX2D | |
Rabbit | |
Affinity Purified | |
RUO | |
644524 | |
Human | |
IgG |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
PBS (pH 7.2), 40% Glycerol with 0.02% Sodium Azide | |
NKX2-4 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:RYSSISRFMGPSAGVNVAGMGSLTGIADAAKSLGPL | |
25 μL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction