Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Nkx2.4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5101641
Description
Antibody detects endogenous levels of total NKX24.
NKX2-4(NK2 Homeobox4) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene includesequence-specific DNA binding and may have transcription factor function.
Specifications
Nkx2.4 | |
Polyclonal | |
Unconjugated | |
NKX2-4 | |
1700001P03Rik; Homeobox protein NK-2 homolog D; homeobox protein Nkx-2.4; LOW QUALITY PROTEIN: homeobox protein Nkx-2.4; NK2 homeobox 1a; NK2 homeobox 4; NK2 transcription factor homolog D; NK2 transcription factor related, locus 4; NKX2.4; Nkx-2.4; NKX2-4; NKX2D; tinman | |
Rabbit | |
Affinity chromatography | |
RUO | |
644524 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9H2Z4 | |
NKX2-4 | |
A synthesized peptide derived from human NKX2-4(Accession Q9H2Z4), corresponding to amino acid residues T92-D142. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction