Learn More
Invitrogen™ Nkx2.4 Polyclonal Antibody

Description
The antibody detects endogenous levels of total NKX2-4 protein.
Specifications
Specifications
| Antigen | Nkx2.4 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | NKX2-4 |
| Gene Accession No. | Q9H2Z4 |
| Gene Alias | 1700001P03Rik; Homeobox protein NK-2 homolog D; homeobox protein Nkx-2.4; LOW QUALITY PROTEIN: homeobox protein Nkx-2.4; NK2 homeobox 1a; NK2 homeobox 4; NK2 transcription factor homolog D; NK2 transcription factor related, locus 4; NKX2.4; Nkx-2.4; NKX2-4; NKX2D; tinman |
| Gene Symbols | NKX2-4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.