Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Nkx2.4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA572762
Description
Nkx2.4 Polyclonal Antibody for Western Blot, ICC/IF, ELISA
NKX2-4(NK2 Homeobox4) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene includesequence-specific DNA binding and may have transcription factor function.
Specifications
Nkx2.4 | |
Polyclonal | |
Unconjugated | |
NKX2-4 | |
1700001P03Rik; Homeobox protein NK-2 homolog D; homeobox protein Nkx-2.4; LOW QUALITY PROTEIN: homeobox protein Nkx-2.4; NK2 homeobox 1a; NK2 homeobox 4; NK2 transcription factor homolog D; NK2 transcription factor related, locus 4; NKX2.4; Nkx-2.4; NKX2-4; NKX2D; tinman | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
228731, 644524 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q9EQM3, Q9H2Z4 | |
NKX2-4 | |
NKX2-4 antibody for an 18 amino acid synthetic peptide near the center of human NKX2-4. The immunogen is within amino acids 220 - 270 of NKX2-4. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction