Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Nkx2.5 Monoclonal Antibody (2E1)
GREENER_CHOICE

Catalog No. PIMA515551
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA515551 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA515551 Supplier Invitrogen™ Supplier No. MA515551
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-15551 targets NKX2.5 in indirect ELISA and WB applications, this antibody also shows reactivity with Human samples. The MA5-15551 immunogen is purified recombinant fragment of human NKX2.5 expressed in E. Coli. MA5-15551 detects NKX2.5 which has a predicted molecular weight of approximately 35kDa.

This gene encodes a homeobox-containing transcription factor. This transcription factor functions in heart formation and development. Mutations in this gene cause atrial septal defect with atrioventricular conduction defect, and also tetralogy of Fallot, which are both heart malformation diseases. Mutations in this gene can also cause congenital hypothyroidism non-goitrous type 5, a non-autoimmune condition. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nkx2.5
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2E1
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation ascites with 0.03% sodium azide
Gene Nkx2-5
Gene Accession No. P52952
Gene Alias Cardiac-specific homeobox; cardiac-specific homeobox 1; CHNG5; CSX; CSX1; Drosophila NK2 transcription factor related, locus 5; FLJ52202; FLJ97166; FLJ97195; FLJ97197; FLJ99536; HLHS2; Homeobox protein CSX; homeobox protein NK-2 homolog E; homeobox protein NKX 2-5; homeobox protein Nkx-2.5; NK2 homeobox 5; NK2 transcription factor related locus 5; NK2 transcription factor related, locus 5; NKX 2-5; NKX2.5; Nkx-2.5; Nkx-25; NKX2-5; NKX2E; NKX4-1; rNKx-2.5; tinman; tinman paralog; VSD3
Gene Symbols Nkx2-5
Host Species Mouse
Immunogen Purified recombinant fragment of human NKX2.5 expressed in E. Coli.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1482
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.