Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Nkx2.5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA585768
Description
Keep as concentrated solution. Predicted reactivity: Human (80%), Mouse (100%), Rat (92%), Dog (81%), Bovine (81%). Positive Control: mouse heart. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a homeobox-containing transcription factor. This transcription factor functions in heart formation and development. Mutations in this gene cause atrial septal defect with atrioventricular conduction defect, and also tetralogy of Fallot, which are both heart malformation diseases. Mutations in this gene can also cause congenital hypothyroidism non-goitrous type 5, a non-autoimmune condition. Alternative splicing results in multiple transcript variants.
Specifications
Nkx2.5 | |
Polyclonal | |
Unconjugated | |
Nkx2-5 | |
Cardiac-specific homeobox; cardiac-specific homeobox 1; CHNG5; CSX; CSX1; Drosophila NK2 transcription factor related, locus 5; FLJ52202; FLJ97166; FLJ97195; FLJ97197; FLJ99536; HLHS2; Homeobox protein CSX; homeobox protein NK-2 homolog E; homeobox protein NKX 2-5; homeobox protein Nkx-2.5; NK2 homeobox 5; NK2 transcription factor related locus 5; NK2 transcription factor related, locus 5; NKX 2-5; NKX2.5; Nkx-2.5; Nkx-25; NKX2-5; NKX2E; NKX4-1; rNKx-2.5; tinman; tinman paralog; VSD3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
114109, 1482, 18091 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.9 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
O35767, P42582, P52952 | |
Nkx2-5 | |
Recombinant protein encompassing a sequence within the center region of mouse Nkx2.5. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction