Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NKX2.6 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP241161
Description
NKX2.6 Polyclonal specifically detects NKX2.6 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.Specifications
NKX2.6 | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
NKX2-6 | |
Antibody was raised against a 15 amino acid synthetic peptide near the center of human NKX2-6. The immunogen is located within amino acids 170 - 220 of NKX2-6. Amino Acid Squence: KRQRQDKSLELAGH | |
0.1 mg | |
Primary | |
NKX2-6 antibody is predicted to not cross-react with other NK2 homeobox family members. At least two isoforms of NKX2-6 are known to exist; this antibody will detect both isoforms | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
Western Blot 1 ug/ml, ELISA, Immunohistochemistry 5 ug/ml, Immunocytochemistry/Immunofluorescence 5 ug/mL, Immunohistochemistry-Paraffin 5 ug/ml | |
CSX2, NK2 homeobox 6, NKX2F, NKX4-2 | |
Rabbit | |
Affinity Purified | |
RUO | |
137814 | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction