Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Nkx2.6 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA550047

Catalog No. PA550047


Only null left
Add to Cart

Description

Description

The antibody detects endogenous levels of total NKX2-6 protein.

Members of the NK-2 family of homeodomain proteins are key regulators of growth and development in several tissues, including brain, heart and pancreas.Nkx-2.5, also designated cardiac specific homeobox protein (Csx), is a homolog of the Drosophila tinman protein and is essential for normal cardiovascular development. Expression of Nkx-2.5 during cardiomyogenesis is required for cardiac septation, in which a single atrium and ventricle are separated into four chambers. Nkx-2.5 binds to DNA as a monomer, a homodimer or as a heterodimer with Nkx-2.3 or Nkx-2.6, which suggests that the specific protein-protein interactions of Nkx-2.5 are involved in its transcriptional regulatory function. Nkx-2.6, also a homolog of the Drosophila tinman protein, is expressed in the caudal pharyngeal pouches,the caudal heart progenitors, the sinus venosus, the outflow tract of the heart and in a short segment of the gut between stages E8.5 and E10.5 of embryogenesis. Expression of Nkx-2.6 overlaps with that of Nkx-2.5 in the pharynx and heart. However, Nkx-2.6 mutant mice are viable and fertile, which suggests that Nkx-2.6 plays a compensatory function to Nkx-2.5.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Nkx2.6
Polyclonal
Unconjugated
NKX2-6
AV044928; CSX2; CTHM; Drosophila NK2 transcription factor related, locus 6; homeobox protein NK2 homolog F; homeobox protein NK-2 homolog F; homeobox protein NKX2.6; homeobox protein Nkx-2.6; NK2 homeobox 6; NK2 transcription factor related, locus 6; Nkx2.6; Nkx-2.6; Nkx2-6; Nkx2f; NKX4-2; tinman; tinman paralog; Tix
Rabbit
Antigen affinity chromatography
RUO
137814
-20°C
Liquid
Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
A6NCS4
NKX2-6
Synthesized peptide derived from internal of human NKX26.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.