Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NKX2.8 Antibody (NKX28/2547), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308561AF700
Description
NKX2.8 Monoclonal antibody specifically detects NKX2.8 in Human samples. It is validated for Western BlotSpecifications
NKX2.8 | |
Monoclonal | |
Alexa Fluor 700 | |
Homeobox protein NK-2 homolog H, homeobox protein Nkx-2.8, NK2 homeobox 8, NK-2 homolog H (Drosophila), NK2 transcription factor related, locus 8, NK2 transcription factor related, locus 8 (Drosophila), NKX-2.8, NKX2.8NK-2 homolog 8, Nkx2-9, NKX2G, NKX2HNK-2 homolog H | |
Human recombinant NKX2.8 protein fragment (around aa 10-123) (exact sequence is proprietary) (Uniprot: O15522) | |
100 μg | |
Primary | |
Human | |
Purified |
Western Blot | |
NKX28/2547 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
26257 | |
Store at 4°C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction