Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Nkx2.8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564071
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564071 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564071 Supplier Invitrogen™ Supplier No. PA564071
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TSGRLSFTVR SLLDLPEQDA QHLPRREPEP RAPQPDPCAA WLDSERGHYP SSDESSLETS PPDSSQR Highest antigen sequence identity to the following orthologs: Mouse - 67%, Rat - 69%.

A murine NKX2-8 was isolated from the Hepal-6 cell line and showed oligonucleotide binding competitive with fetoprotein transcription factor. Nkx2.8 bound to the active AFP promoter, and antisense inhibition of Nkx2.8 mRNA translation selectively reduced expression of both the endogenous human AFP gene and transfected reporters containing the rat AFP promoter.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nkx2.8
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NKX2-8
Gene Accession No. O15522
Gene Alias Drosophila NK2 transcription factor related, locus 9; homeobox protein NK-2 homolog H; Homeobox protein Nkx-2.8; homeobox protein Nkx-2.9; NK2 homeobox 8; NK2 homeobox 9; NK-2 homolog 8; NK-2 homolog H; NK2 transcription factor related, locus 8; NK2 transcription factor related, locus 9; NKX2.8; NKX-2.8; Nkx2.9; Nkx-2.9; Nkx2-8; Nkx2-9; NKX2G; NKX2H; tinman
Gene Symbols NKX2-8
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Nkx2.8. Recombinant protein control fragment (Product #RP-100065).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26257
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.