Learn More
NKX2 Mouse anti-Human, Clone: OTI15A10, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF803319

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a protein initially identified as a thyroid-specific transcription factor. The encoded protein binds to thyroglobulin promoter and regulates the expression of thyroid-specific genes but has also been shown to regulate the expression of genes involved in morphogenesis. Mutations and deletions in this gene are associated with benign hereditary chorea, choreoathetosis, congenital hypothyroidism, and neonatal respiratory distress, and may be associated with thyroid cancer. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
NKX2 | |
Monoclonal | |
Unconjugated | |
NKX2-1 | |
BCH, BHC, NK-2, NKX2.1, NKX2A, NMTC1, T/EBP, TEBP, TITF1, TTF-1, TTF1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
7080 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI15A10 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P43699 | |
NKX2-1 | |
Full length human recombit protein of human NKX2-1 ( NP_003308) produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.