Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NKX3.1 Antibody (NKX3.1/3350), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308572C
Description
NKX3.1 Monoclonal antibody specifically detects NKX3.1 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin), Protein ArraySpecifications
NKX3.1 | |
Monoclonal | |
DyLight 650 | |
BAPX2homeobox protein Nkx-3.1, Homeobox protein NK-3 homolog A, NK homeobox (Drosophila), family 3, A, NK3 homeobox 1, NK3 transcription factor homolog A, NK3 transcription factor related, locus 1, NK3 transcription factor related, locus 1 (Drosophila), NKX3.1NKX3, NKX3ANK homeobox, family 3, A | |
Recombinant fragment (around aa 92-224) of human NKX3.1 protein (exact sequence is proprietary) (Uniprot: Q99801) | |
100 μg | |
Cancer, Chromatin Research, Prostate Cancer, Transcription Factors and Regulators | |
4824 | |
Store at 4°C in the dark. | |
IgG2b κ |
ELISA, Immunohistochemistry (Paraffin), Protein Array | |
NKX3.1/3350 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction