Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NKX3.1 Mouse anti-Human, Clone: rNKX3.1/6620, Novus Biologicals™

Mouse Monoclonal Antibody
$268.00
Specifications
| Antigen | NKX3.1 |
|---|---|
| Clone | rNKX3.1/6620 |
| Concentration | 0.2 mg/ml |
| Dilution | ELISA, Immunohistochemistry-Paraffin 1-2 ug/ml |
| Applications | ELISA, Immunohistochemistry (Paraffin) |
Description
NKX3.1 Monoclonal specifically detects NKX3.1 in Human samples. It is validated for ELISA, Immunohistochemistry-Paraffin.Specifications
| NKX3.1 | |
| 0.2 mg/ml | |
| ELISA, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cancer, Chromatin Research, Prostate Cancer, Transcription Factors and Regulators | |
| 10 mM PBS with 0.05% BSA | |
| 4824 | |
| IgG1 κ | |
| Protein A or G purified |
| rNKX3.1/6620 | |
| ELISA, Immunohistochemistry-Paraffin 1-2 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| BAPX2homeobox protein Nkx-3.1, Homeobox protein NK-3 homolog A, NK homeobox (Drosophila), family 3, A, NK3 homeobox 1, NK3 transcription factor homolog A, NK3 transcription factor related, locus 1, NK3 transcription factor related, locus 1 (Drosophila), NKX3.1NKX3, NKX3ANK homeobox, family 3, A | |
| Recombinant fragment (around aa 92-224) of human NKX3.1 protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title