Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Nkx3.1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA585062
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA585062 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA585062 Supplier Invitrogen™ Supplier No. PA585062
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MLRVPEPRPG EAKAEGAAPP TPSKPLTSFL IQDILRDG Highest antigen sequence indentity to the following orthologs: Rat - 64%, Mouse - 67%.

The homeodomain-containing transcription factor NKX3-1 is a putative prostate tumor suppressor that is expressed in a largely prostate-specific and androgen-regulated manner. Loss of NKX3-1 protein expression is a common finding in human prostate carcinomas and prostatic intraepithelial neoplasia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nkx3.1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NKX3-1
Gene Accession No. Q99801
Gene Alias bagpipe; BAPX2; Bax; Drosophila NK-3 transcription factor, locus 1; homeobox protein NK-3 homolog A; Homeobox protein Nkx-3.1; NK homeobox, family 3, A; NK3 homeobox 1; NK3 transcription factor homolog A; NK3 transcription factor related, locus 1; NK-3 transcription factor, locus 1; NK-3 transcription factor, locus 1 (Drosophila); NKX3; NKX3.1; Nkx-3.1; Nkx3-1; NKX3-1 antibody; NKX3A; prostate-specific ag
Gene Symbols NKX3-1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NKX3-1. Recombinant protein control fragment (Product #RP-108300).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4824
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.