Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Nkx3.1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA580856
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for human NKX3-1.
The homeodomain-containing transcription factor NKX3-1 is a putative prostate tumor suppressor that is expressed in a largely prostate-specific and androgen-regulated manner. Loss of NKX3-1 protein expression is a common finding in human prostate carcinomas and prostatic intraepithelial neoplasia.
Specifications
Nkx3.1 | |
Polyclonal | |
Unconjugated | |
NKX3-1 | |
bagpipe; BAPX2; Bax; Drosophila NK-3 transcription factor, locus 1; homeobox protein NK-3 homolog A; Homeobox protein Nkx-3.1; NK homeobox, family 3, A; NK3 homeobox 1; NK3 transcription factor homolog A; NK3 transcription factor related, locus 1; NK-3 transcription factor, locus 1; NK-3 transcription factor, locus 1 (Drosophila); NKX3; NKX3.1; Nkx-3.1; Nkx3-1; NKX3-1 antibody; NKX3A; prostate-specific ag | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
4824 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with no preservative | |
Q99801 | |
NKX3-1 | |
A synthetic peptide corresponding to the N-terminus of the human NKX3-1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction