Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NKX3.1 Rabbit anti-Human, Clone: RM430, Novus Biologicals™
SDP

Rabbit Monoclonal Antibody

$181.90 - $548.00

Specifications

Antigen NKX3.1
Clone RM430
Dilution Immunohistochemistry 1:100-1:400
Applications Immunohistochemistry
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB119708
SDP
View Documents
Novus Biologicals
NBP30918025UL
25 μg
Each for $181.90
Only null left
Add to Cart
 
NB119707
SDP
View Documents
Novus Biologicals
NBP309180100UL
100 μg
Each for $548.00
Only null left
Add to Cart
 
Description

Description

NKX3.1 Monoclonal specifically detects NKX3.1 in Human samples. It is validated for Immunohistochemistry.
Specifications

Specifications

NKX3.1
Immunohistochemistry 1:100-1:400
Monoclonal
Purified
RUO
Human
BAPX2homeobox protein Nkx-3.1, Homeobox protein NK-3 homolog A, NK homeobox (Drosophila), family 3, A, NK3 homeobox 1, NK3 transcription factor homolog A, NK3 transcription factor related, locus 1, NK3 transcription factor related, locus 1 (Drosophila), NKX3.1NKX3, NKX3ANK homeobox, family 3, A
A peptide corresponding to the N-terminus of human NKX3.1
Primary
Store at -20°C.
RM430
Immunohistochemistry
Unconjugated
Rabbit
Cancer, Chromatin Research, Prostate Cancer, Transcription Factors and Regulators
PBS, 50% Glycerol, 1% BSA
4824
IgG
Protein A purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.